Gene Transcripts

Gene Symbol
Hspa12a
Entrez Gene ID
73442
Gene Name
heat shock protein 12A
Gene Aliases
1700063D12Rik, AI118035, AI840429, AW048913, AW556406, D5Mgi40, mKIAA0417
Chromosome Location
Chr. 19: 58795750 - 58943545 on Build GRCm38
UniGene ID
Mm.39739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175199.3 NP_780408.1 5 597
GenBank mRNA AK006867.1 - 5 581
AK038491.1 - 5 569
AK087732.1 - 6 755
AK141925.1 - 5 598
AK165073.1 - 5 625
AY196789.1 - 5 596
BC030362.1 - 5 567 5369051

View Full Product Details