Gene Transcripts
- Gene Symbol
- Arsg
- Entrez Gene ID
- 74008
- Gene Name
- arylsulfatase G
- Gene Aliases
- 6330406P08Rik, AI846872
- Chromosome Location
- Chr. 11: 109450845 - 109573330 on Build GRCm38
- UniGene ID
- Mm.482224
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166177.1 |
NP_001159649.1
|
2, 3 |
220 |
|
NM_028710.3 |
NP_082986.3
|
11, 12 |
1947 |
|
GenBank mRNA |
AK018132.2 |
- |
8, 9 |
1364 |
|
AK158726.1 |
- |
11, 12 |
1948 |
|
AK173082.1 |
- |
8, 9 |
1046 |
|
AK186315.1 |
- |
3, 4 |
228 |
|
BC022158.1 |
- |
7, 8 |
777 |
|
BC039629.1 |
- |
2, 3 |
177 |
|
BC084731.1 |
- |
8, 9 |
1637 |
6813261 |
BY095625.1 |
- |
2, 3 |
202 |
|
View Full Product Details