Gene Transcripts

Gene Symbol
Armc8
Entrez Gene ID
74125
Gene Name
armadillo repeat containing 8
Gene Aliases
1200015K23Rik, Gid5, HSPC056
Chromosome Location
Chr. 9: 99478385 - 99568936 on Build GRCm38
UniGene ID
Mm.234823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028768.3 NP_083044.2 19, 20 2082
GenBank mRNA AK004793.2 - 19, 20 2083
BC030311.1 - 1, 2 234

View Full Product Details