Gene Transcripts

Gene Symbol
NMB
Entrez Gene ID
4828
Gene Name
neuromedin B
Gene Aliases
Chromosome Location
Chr. 15: 84655129 - 84658611 on Build GRCh38
UniGene ID
Hs.386470
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021077.3 NP_066563.2 3 968
NM_205858.1 NP_995580.1 3 963
GenBank mRNA BC007407.2 - 3 956
BC007431.2 - 3 956 3030430
BC008603.1 - 3 588 4185578
M21551.1 - 3 586

View Full Product Details