Gene Transcripts

Gene Symbol
Klra19
Entrez Gene ID
93971
Gene Name
killer cell lectin-like receptor, subfamily A, member 19
Gene Aliases
Ly49s
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.14112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053154.2 NP_444384.1 3 269
GenBank mRNA AF288378.1 - 3 269
BC116822.1 - 3 228 40126141
BC116824.1 - 3 222 40126143

View Full Product Details