Gene Transcripts

Gene Symbol
Mex3b
Entrez Gene ID
108797
Gene Name
mex3 homolog B (C. elegans)
Gene Aliases
4931439A04Rik, M-04814, Rkhd3, mKIAA2009
Chromosome Location
Chr. 7: 82867333 - 82871576 on Build GRCm38
UniGene ID
Mm.306789
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175366.3 NP_780575.2 2 892
GenBank mRNA AK173330.1 - 1 1705
BC108401.1 - 1 100

View Full Product Details