Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001348.2 | NP_001339.1 | 7 | 1019 | |
XM_005259508.1 | XP_005259565.1 | 7 | 894 | ||
GenBank mRNA | AB007144.1 | - | 6 | 815 | |
AB022341.1 | - | 6 | 767 | ||
AB265224.1 | - | 4 | 232 | ||
AK027590.1 | - | 7 | 936 | ||
AK074799.1 | - | 6 | 791 | ||
AK093792.1 | - | 1 | 772 | ||
AK097643.1 | - | 7 | 830 | ||
BC126430.1 | - | 6 | 758 | 8992146 | |
BC126432.1 | - | 6 | 756 | 8992148 |