Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008565.3 | NP_032591.3 | 4 | 631 | |
GenBank mRNA | AK011743.1 | - | 4 | 566 | |
AK036653.1 | - | 4 | 591 | ||
AK083618.1 | - | 4 | 545 | ||
AK088796.1 | - | 4 | 551 | ||
AK089999.1 | - | 4 | 535 | ||
AK135422.1 | - | 4 | 533 | ||
AK141132.1 | - | 4 | 597 | ||
AK141644.1 | - | 4 | 632 | ||
AK146662.1 | - | 4 | 554 | ||
AK151499.1 | - | 4 | 545 | ||
AK157725.1 | - | 4 | 533 | ||
AK220442.1 | - | 4 | 532 | ||
BC013094.1 | - | 4 | 587 | 3584640 | |
D26089.1 | - | 4 | 537 |