Gene Transcripts

Gene Symbol
Hist1h1d
Entrez Gene ID
14957
Gene Name
histone cluster 1, H1d
Gene Aliases
H1.3, H1D, H1f3, H1s-4
Chromosome Location
Chr. 13: 23555032 - 23555807 on Build GRCm38
UniGene ID
Mm.247046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK054269.1 - 922
AK155411.1 - 926
BC117045.1 - 1 921 40126364
BC119173.1 - 1 921 8733922

View Full Product Details