Gene Transcripts

Gene Symbol
Ddx5
Entrez Gene ID
13207
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 5
Gene Aliases
2600009A06Rik, G17P1, HUMP68, Hlr1, p68
Chromosome Location
Chr. 11: 106780355 - 106789268 on Build GRCm38
UniGene ID
Mm.491096
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007840.3 NP_031866.2 13 1953
GenBank mRNA AK013106.1 - 6 842
AK044792.1 - 13 1954
AK076018.1 - 13 1963
AK088887.1 - 13 1954
AK129022.1 - 12 3227
AK132779.1 - 13 2000
AK135173.1 - 13 1954
AK166045.1 - 13 1954
AK166698.1 - 14 2086
AK184106.1 - 2 393
BC009142.1 - 2 1266
BC046554.1 - 13 1931
BC062916.1 - 13 1938 6827275
BC086320.1 - 13 1876
BC108369.1 - 13 1798 6312791
BC129873.1 - 13 1831 40110366
BC129874.1 - 13 1834 40110367
X65627.1 - 13 1935

View Full Product Details