Gene Transcripts

Gene Symbol
Gm4763
Entrez Gene ID
210155
Gene Name
predicted gene 4763
Gene Aliases
EG210155
Chromosome Location
Chr. 7: 24714591 - 24726934 on Build GRCm38
UniGene ID
Mm.425399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177593.1 NP_808261.1 2 147
GenBank mRNA BC037749.1 - 2 147

View Full Product Details