Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175141.5 | NP_780350.1 | 1 | 113 | |
GenBank mRNA | AK078166.1 | - | 1 | 114 | |
AK146041.1 | - | 1 | 135 | ||
AK169085.1 | - | 1 | 120 | ||
BC038642.1 | - | 1 | 101 | ||
BC051146.1 | - | 1 | 125 | ||
BC137660.1 | - | 1 | 206 | 8860679 |