Gene Transcripts

Gene Symbol
Serpind1
Entrez Gene ID
79224
Gene Name
serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Gene Aliases
rls2var1
Chromosome Location
Chr. 11: 87913814 - 87924880 on Build Rnor_6.0
UniGene ID
Rn.10553
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024382.1 NP_077358.1 5 1261
XM_006248709.2 XP_006248771.1 4 1268
GenBank mRNA BC091145.1 - 5 1241
FQ210385.1 - 2 294
FQ218607.1 - 4 1238
X74549.1 - 5 1261
X74550.1 - 4 1208

View Full Product Details