Gene Transcripts
- Gene Symbol
- SCN8A
- Entrez Gene ID
- 6334
- Gene Name
- sodium voltage-gated channel alpha subunit 8
- Gene Aliases
- CERIII, CIAT, EIEE13, MED, NaCh6, Nav1.6, PN4
- Chromosome Location
- Chr. 12: 51589959 - 51812865 on Build GRCh38
- UniGene ID
- Hs.436550
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177984.2 |
NP_001171455.1
|
2, 3 |
455 |
|
NM_014191.3 |
NP_055006.1
|
2, 3 |
455 |
|
XM_006719556.3 |
XP_006719619.1
|
2, 3 |
1046 |
|
XM_011538650.2 |
XP_011536952.1
|
2, 3 |
474 |
|
XM_011538651.2 |
XP_011536953.1
|
2, 3 |
439 |
|
XM_017019794.1 |
XP_016875283.1
|
2, 3 |
452 |
|
XM_017019795.1 |
XP_016875284.1
|
2, 3 |
471 |
|
XM_017019796.1 |
XP_016875285.1
|
2, 3 |
471 |
|
GenBank mRNA |
AB027567.1 |
- |
2, 3 |
455 |
|
AF225988.1 |
- |
1, 2 |
277 |
|
AK309213.1 |
- |
2, 3 |
405 |
|
FJ611941.1 |
- |
1, 2 |
277 |
|
View Full Product Details