Gene Transcripts
- Gene Symbol
- TMEM132B
- Entrez Gene ID
- 114795
- Gene Name
- transmembrane protein 132B
- Gene Aliases
- Chromosome Location
- Chr. 12: 125186750 - 125662377 on Build GRCh38
- UniGene ID
- Hs.524838
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB058689.1 |
- |
4 |
4547 |
|
View Full Product Details