Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000238.3 | NP_000229.1 | 7 | 2185 | |
NM_001204798.1 | NP_001191727.1 | 3 | 1090 | ||
NM_172056.2 | NP_742053.1 | 7 | 2185 | ||
NM_172057.2 | NP_742054.1 | 3 | 1090 | ||
XM_011516185.2 | XP_011514487.1 | 6 | 1596 | ||
XM_011516186.2 | XP_011514488.1 | 7 | 2177 | ||
XM_017012195.1 | XP_016867684.1 | 6 | 2266 | ||
XM_017012196.1 | XP_016867685.1 | 7 | 1726 | ||
GenBank mRNA | AB044806.1 | - | 7 | 1967 | |
AF363636.1 | - | 7 | 1784 | ||
AJ512214.1 | - | 3 | 781 | ||
AJ609614.1 | - | 3 | 764 | ||
AY927502.1 | - | 1 | 31 | ||
BC001914.1 | - | 6 | 1441 | ||
BC127673.1 | - | 3 | 1079 | ||
BT007336.1 | - | 6 | 1436 | ||
DQ525913.1 | - | 7 | 2185 | ||
FJ938021.1 | - | 5 | 2617 | ||
U04270.1 | - | 7 | 1967 |