Gene Transcripts

Gene Symbol
Gm13157
Entrez Gene ID
100041677
Gene Name
predicted gene 13157
Gene Aliases
OTTMUSG00000010679
Chromosome Location
Chr. 4: 147753974 - 147810843 on Build GRCm38
UniGene ID
Mm.327194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127189.3 NP_001120661.1 2 145
GenBank mRNA AI789447.1 - 2 145
AK163132.1 - 2 198

View Full Product Details