Gene Transcripts

Gene Symbol
2010016I18Rik
Entrez Gene ID
69206
Gene Name
RIKEN cDNA 2010016I18 gene
Gene Aliases
Chromosome Location
Chr. 3: 106481986 - 106485913 on Build GRCm38
UniGene ID
Mm.14816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK040034.1 - 1 272
AK079963.1 - 1 656
AK080971.1 - 2 362
AK140363.1 - 2 362

View Full Product Details