Gene Transcripts

Gene Symbol
Psmd10
Entrez Gene ID
116722
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 10
Gene Aliases
p28Gank
Chromosome Location
Chr. X: 112323188 - 112328642 on Build Rnor_6.0
UniGene ID
Rn.116732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053925.1 NP_446377.1 591
GenBank mRNA AB022014.1 - 591
BC158584.1 - 434 8369120

View Full Product Details