Gene Transcripts

Gene Symbol
Impg2
Entrez Gene ID
245919
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
Pg10.2
Chromosome Location
Chr. 11: 46615168 - 46694553 on Build Rnor_6.0
UniGene ID
Rn.10612
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139088.2 NP_620788.2 2 352
XM_008768628.1 XP_008766850.1 2 1340
GenBank mRNA U76717.1 - 2 352

View Full Product Details