Gene Transcripts

Gene Symbol
Cd2ap
Entrez Gene ID
316258
Gene Name
CD2-associated protein
Gene Aliases
CMS
Chromosome Location
Chr. 9: 20794680 - 20858438 on Build Rnor_6.0
UniGene ID
Rn.212220
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181475.2 NP_852140.1 1 111
GenBank mRNA AY114147.1 - 1 107
AY205155.1 - 1 111
CB545525.1 - 231

View Full Product Details