Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012671.2 | NP_036803.1 | 3 | 267 | |
XM_006236822.2 | XP_006236884.1 | 3 | 1160 | ||
XM_008763049.1 | XP_008761271.1 | 3 | 1164 | ||
XM_008763050.1 | XP_008761272.1 | 3 | 1161 | ||
GenBank mRNA | KF366251.1 | - | 3 | 123 | |
M31076.1 | - | 3 | 267 | ||
X02004.1 | - | 4 | 267 | ||
X61485.1 | - | 2 | 48 | ||
X61486.1 | - | 2 | 48 | ||
X61487.1 | - | 2 | 48 |