Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026048.4 | NP_080324.1 | 2 | 153 | |
NM_027223.1 | NP_081499.1 | 2 | 153 | ||
GenBank mRNA | AK004355.1 | - | 2 | 108 | |
AK007812.1 | - | 2 | 143 | ||
AK032155.1 | - | 2 | 142 | ||
AK076126.1 | - | 2 | 123 | ||
AK160662.1 | - | 2 | 123 | ||
BC011066.1 | - | 2 | 123 | 3985202 | |
BC021867.1 | - | 2 | 105 | 3710025 |