Gene Transcripts

Gene Symbol
Olfr923
Entrez Gene ID
258812
Gene Name
olfactory receptor 923
Gene Aliases
MOR164-2
Chromosome Location
Chr. 9: 38827615 - 38828682 on Build GRCm38
UniGene ID
Mm.377702
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146816.2 NP_667027.2 1 805
GenBank mRNA BC132247.1 - 1 805
BC132249.1 - 1 805 40130526

View Full Product Details