gRNA Design Details
- Target DNA Sequence
- ATTACCTTCCCACCCAGCAG
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 118352565 - 118352587 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS12
- Entrez Gene ID
-
8492
- Gene Name
- protease, serine 12
- Gene Aliases
-
BSSP-3, BSSP3, MRT1
- Chromosome Location
-
Chr.4: 118280038 - 118353042 on GRCh38
- UniGene ID
- Hs.445857
- Species
-
Human