gRNA Design Details
- Target DNA Sequence
- TCTGACACTGTTTGGCAGTT
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 164700210 - 164700232 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint4
- Entrez Gene ID
-
78239
- Gene Name
- serine protease inhibitor, Kunitz type 4
- Gene Aliases
-
9230105I15Rik
- Chromosome Location
-
Chr.2: 164698501 - 164702448 on GRCm38
- UniGene ID
- Mm.160060
- Species
-
Mouse