gRNA Design Details
- Target DNA Sequence
- CTGAGCATTACGTGCTGGAA
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 19768458 - 19768480 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bcam
- Entrez Gene ID
-
57278
- Gene Name
- basal cell adhesion molecule
- Gene Aliases
-
1200005K12Rik, B-CAM, Gplu, Lu
- Chromosome Location
-
Chr.7: 19756138 - 19770532 on GRCm38
- UniGene ID
- Mm.29236
- Species
-
Mouse