gRNA Design Details
- Target DNA Sequence
- GTGGCTTTGGTTCTGTAACA
- PAM Sequence
- GGG
- Application
- Gene Knockout
Score:
77.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rbbp7
- Entrez Gene ID
-
245688
- Gene Name
- retinoblastoma binding protein 7
- Gene Aliases
-
AA409861, AI173248, AU019541, BB114024, mRbAp46
- Chromosome Location
-
Chr.X: 162760372 - 162779090 on GRCm38
- UniGene ID
- Mm.270186
- Species
-
Mouse