gRNA Design Details

Target DNA Sequence
GGCCCAGTCTTACCGTTCAG
PAM Sequence
TGG
Application
Gene Knockout
Score: 71.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
RBBP7
Entrez Gene ID
5931
Gene Name
RB binding protein 7, chromatin remodeling factor
Gene Aliases
RbAp46
Chromosome Location
Chr.X: 16844650 - 16870411 on GRCh38
UniGene ID
Hs.495755
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001198719.1 NP_001185648.1 2
NM_002893.3 NP_002884.1 2