gRNA Design Details
- Target DNA Sequence
- CGAAGTCAACTCTGCGAAAG
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 164715441 - 164715463 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint5
- Entrez Gene ID
-
641368
- Gene Name
- serine protease inhibitor, Kunitz type 5
- Gene Aliases
-
Gm14316, OTTMUSG00000016293
- Chromosome Location
-
Chr.2: 164715305 - 164718068 on GRCm38
- UniGene ID
- Mm.422380
- Species
-
Mouse