gRNA Design Details
- Target DNA Sequence
- CCCGGTAATGTTTGGCTAGT
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 38450162 - 38450184 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adprh
- Entrez Gene ID
-
11544
- Gene Name
- ADP-ribosylarginine hydrolase
- Gene Aliases
-
Arh1
- Chromosome Location
-
Chr.16: 38445399 - 38452689 on GRCm38
- UniGene ID
- Mm.20047
- Species
-
Mouse