gRNA Design Details
- Target DNA Sequence
- CACTATGAGATGGACCCGGG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 28675340 - 28675362 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.8/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Opcml
- Entrez Gene ID
-
330908
- Gene Name
- opioid binding protein/cell adhesion molecule-like
- Gene Aliases
-
2900075O15Rik, 3732419F12, AI844366, B930023M13Rik, C230027C17, Gm181, Obcam
- Chromosome Location
-
Chr.9: 27790952 - 28925410 on GRCm38
- UniGene ID
- Mm.379474
- Species
-
Mouse