gRNA Design Details
- Target DNA Sequence
- ACCTCGTCCCAGTGTAGACA
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 123447456 - 123447478 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss12
- Entrez Gene ID
-
19142
- Gene Name
- protease, serine 12 neurotrypsin (motopsin)
- Gene Aliases
-
Bssp-3, motopsin
- Chromosome Location
-
Chr.3: 123446913 - 123506602 on GRCm38
- UniGene ID
- Mm.9431
- Species
-
Mouse