gRNA Design Details

Target DNA Sequence
TGACCAGGATGATCACACGA
PAM Sequence
GGG
Target locus
Chr.11: 132657210 - 132657232 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
OPCML
Entrez Gene ID
4978
Gene Name
opioid binding protein/cell adhesion molecule like
Gene Aliases
IGLON1, OBCAM, OPCM
Chromosome Location
Chr.11: 132403361 - 133532983 on GRCh38
UniGene ID
Hs.4817
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001012393.2 NP_001012393.1 3
NM_001319103.1 NP_001306032.1 2
NM_001319104.1 NP_001306033.1 2
NM_001319105.1 NP_001306034.1 3
NM_001319106.1 NP_001306035.1 2
NM_002545.4 NP_002536.1 2
XM_006718846.2 XP_006718909.1 3
XM_011542856.2 XP_011541158.1 4