gRNA Design Details
- Target DNA Sequence
- GTTTGGGACAACCACCGAAC
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 16236701 - 16236723 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
97.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RSG1
- Entrez Gene ID
-
79363
- Gene Name
- REM2 and RAB like small GTPase 1
- Gene Aliases
-
C1orf89
- Chromosome Location
-
Chr.1: 16231687 - 16237445 on GRCh38
- UniGene ID
- Hs.546430
- Species
-
Human