gRNA Design Details

Target DNA Sequence
TCTTGCGCAGAATGCAAGCC
PAM Sequence
AGG
Target locus
Chr.1: 16236649 - 16236671 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 89.19/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
RSG1
Entrez Gene ID
79363
Gene Name
REM2 and RAB like small GTPase 1
Gene Aliases
C1orf89
Chromosome Location
Chr.1: 16231687 - 16237445 on GRCh38
UniGene ID
Hs.546430
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_030907.3 NP_112169.2 1
XM_011542126.2 XP_011540428.1 1