gRNA Design Details
- Target DNA Sequence
- GTGCATTTGTCTTCCCGACT
- PAM Sequence
- GGG
- Application
- Gene Knockout
Score:
89.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RBBP7
- Entrez Gene ID
-
5931
- Gene Name
- RB binding protein 7, chromatin remodeling factor
- Gene Aliases
-
RbAp46
- Chromosome Location
-
Chr.X: 16844650 - 16870411 on GRCh38
- UniGene ID
- Hs.495755
- Species
-
Human