gRNA Design Details
- Target DNA Sequence
- CTTGGTAATGCTTAGCAAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 119582412 - 119582434 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADPRH
- Entrez Gene ID
-
141
- Gene Name
- ADP-ribosylarginine hydrolase
- Gene Aliases
-
ARH1
- Chromosome Location
-
Chr.3: 119579433 - 119589954 on GRCh38
- UniGene ID
- Hs.99884
- Species
-
Human