gRNA Design Details

Target DNA Sequence
AGAGTCTTCCTGTCGTATTG
PAM Sequence
GGG
Target locus
Chr.7: 142751862 - 142751884 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 86.12/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS1
Entrez Gene ID
5644
Gene Name
protease, serine 1
Gene Aliases
TRP1, TRY1, TRY4, TRYP1
Chromosome Location
Chr.7: 142740235 - 142753076 on GRCh38
UniGene ID
Hs.382212
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002769.4 NP_002760.1 3
XM_011516411.1 XP_011514713.1 5