gRNA Design Details
- Target DNA Sequence
- AGAGTCTTCCTGTCGTATTG
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 142751862 - 142751884 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.12/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS1
- Entrez Gene ID
-
5644
- Gene Name
- protease, serine 1
- Gene Aliases
-
TRP1, TRY1, TRY4, TRYP1
- Chromosome Location
-
Chr.7: 142740235 - 142753076 on GRCh38
- UniGene ID
- Hs.382212
- Species
-
Human