gRNA Design Details

Target DNA Sequence
AGCCAAATAGGTCAAGACCA
PAM Sequence
AGG
Target locus
Chr.1: 150508232 - 150508254 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 86.88/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ECM1
Entrez Gene ID
1893
Gene Name
extracellular matrix protein 1
Gene Aliases
URBWD
Chromosome Location
Chr.1: 150508011 - 150513789 on GRCh38
UniGene ID
Hs.81071
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001202858.1 NP_001189787.1 1
NM_004425.3 NP_004416.2 1
NM_022664.2 NP_073155.2 1