gRNA Design Details
- Target DNA Sequence
- GCAGCTTTTCCTGTTGGAGA
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 150509950 - 150509972 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ECM1
- Entrez Gene ID
-
1893
- Gene Name
- extracellular matrix protein 1
- Gene Aliases
-
URBWD
- Chromosome Location
-
Chr.1: 150508011 - 150513789 on GRCh38
- UniGene ID
- Hs.81071
- Species
-
Human