gRNA Design Details
- Target DNA Sequence
- AGGATTCGGAATACTACGAT
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 26845980 - 26846002 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Abo
- Entrez Gene ID
-
80908
- Gene Name
- ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase)
- Gene Aliases
-
NAGAT
- Chromosome Location
-
Chr.2: 26842496 - 26864995 on GRCm38
- UniGene ID
- Mm.160386
- Species
-
Mouse