gRNA Design Details
- Target DNA Sequence
- TAATGGTCGTGGGTTCCCGT
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 44811305 - 44811327 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
95.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BCAM
- Entrez Gene ID
-
4059
- Gene Name
- basal cell adhesion molecule (Lutheran blood group)
- Gene Aliases
-
AU, CD239, LU, MSK19
- Chromosome Location
-
Chr.19: 44809059 - 44821421 on GRCh38
- UniGene ID
- Hs.625725
- Species
-
Human