Gene Transcripts

Gene Symbol
CACNB2
Entrez Gene ID
783
Gene Name
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases
CACNLB2, CAVB2, MYSB
Chromosome Location
Chr. 10: 18140560 - 18545881 on Build GRCh38
UniGene ID
Hs.59093
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000724.3 NP_000715.2 9 1461
NM_001167945.1 NP_001161417.1 10 1180
NM_201570.2 NP_963864.1 9 1315
NM_201571.3 NP_963865.2 10 1294
NM_201572.3 NP_963866.2 10 1222
NM_201590.2 NP_963884.2 9 1091
NM_201593.2 NP_963887.2 10 957
NM_201596.2 NP_963890.2 10 1071
NM_201597.2 NP_963891.1 10 999
XM_005252588.3 XP_005252645.1 9 1205
XM_005252591.3 XP_005252648.1 4 266
XM_006717502.3 XP_006717565.1 9 1124
XM_011519659.2 XP_011517961.1 9 1019
XM_011519660.2 XP_011517962.1 9 1562
XM_017016625.1 XP_016872114.1 10 1179
GenBank mRNA AB208917.1 - 9 1347
AF137376.1 - 10 1009
AF137377.1 - 9 852
AF285239.2 - 9 849
AF423189.1 - 9 846
AF423190.1 - 10 927
AF423191.1 - 10 1011
AF423192.1 - 9 867
AF465485.1 - 9 741
AK293937.1 - 9 762
AK296471.1 - 10 978
AK299873.1 - 9 1217
AK307526.1 - 10 1180
AK307800.1 - 9 923
AY393858.1 - 10 927
AY393859.1 - 9 849
AY393860.1 - 10 939
AY393861.1 - 10 855
AY393862.1 - 9 777
BC136409.1 - 10 1034 9020397
BM312516.1 - 7 464
S60415.1 - 10 1071
U95019.1 - 9 1347

View Full Product Details