Gene Transcripts
- Gene Symbol
- NUTM2A-AS1
- Entrez Gene ID
- 728190
- Gene Name
- NUTM2A antisense RNA 1
- Gene Aliases
- FAM22A-AS1
- Chromosome Location
- Chr. 10: 87238667 - 87342558 on Build GRCh38
- UniGene ID
- Hs.440648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024397.1 |
|
5 |
562 |
|
GenBank mRNA |
AI832214.1 |
- |
2 |
340 |
|
BE856553.1 |
- |
2 |
260 |
|
View Full Product Details