Gene Transcripts

Gene Symbol
FMR1NB
Entrez Gene ID
158521
Gene Name
fragile X mental retardation 1 neighbor
Gene Aliases
CT37, NY-SAR-35, NYSAR35
Chromosome Location
Chr. X: 147981329 - 148026667 on Build GRCh38
UniGene ID
Hs.128580
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152578.2 NP_689791.1 3 541
GenBank mRNA AK098602.1 - 3 526
AY211917.1 - 3 534
BC034320.1 - 3 541 4836772

View Full Product Details