Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 136115987-136116492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR4
Entrez Gene ID
7852
Gene Name
C-X-C motif chemokine receptor 4
Gene Aliases
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS
UniGene
Hs.593413
Gene Chromosome Location
Chr.2: 136114349-136118155 on Build GRCh38
Transcripts
NM_001008540.1 NM_003467.2

SNP Detail

SNP ID Chromosome Location
rs933409514 Chr.2: 136116258-136116258 on Build GRCh38
rs533230774 Chr.2: 136116388-136116388 on Build GRCh38
rs1046828698 Chr.2: 136116376-136116376 on Build GRCh38
rs564357587 Chr.2: 136116387-136116387 on Build GRCh38
rs561969013 Chr.2: 136116274-136116274 on Build GRCh38
rs189487718 Chr.2: 136116457-136116457 on Build GRCh38
rs1057215998 Chr.2: 136116344-136116344 on Build GRCh38
rs993935019 Chr.2: 136116093-136116093 on Build GRCh38
rs531949370 Chr.2: 136116459-136116459 on Build GRCh38
rs916256092 Chr.2: 136116018-136116018 on Build GRCh38
rs1046318353 Chr.2: 136116058-136116058 on Build GRCh38
rs1043312450 Chr.2: 136116289-136116289 on Build GRCh38
rs573037333 Chr.2: 136116221-136116221 on Build GRCh38
rs996168097 Chr.2: 136116437-136116437 on Build GRCh38
rs947570324 Chr.2: 136116288-136116288 on Build GRCh38
rs542054261 Chr.2: 136116224-136116224 on Build GRCh38
rs1017665362 Chr.2: 136116244-136116244 on Build GRCh38
rs1006306548 Chr.2: 136116236-136116236 on Build GRCh38
rs936989779 Chr.2: 136116333-136116333 on Build GRCh38
rs539571808 Chr.2: 136116147-136116147 on Build GRCh38
rs988917830 Chr.2: 136116272-136116272 on Build GRCh38
rs906112228 Chr.2: 136116299-136116299 on Build GRCh38
rs879110090 Chr.2: 136116073-136116073 on Build GRCh38
rs1025452941 Chr.2: 136116184-136116184 on Build GRCh38
rs975155718 Chr.2: 136116244-136116247 on Build GRCh38
rs868404947 Chr.2: 136116109-136116109 on Build GRCh38
rs553124006 Chr.2: 136116197-136116197 on Build GRCh38
rs544583045 Chr.2: 136116345-136116345 on Build GRCh38
rs546655844 Chr.2: 136116405-136116405 on Build GRCh38
rs76931498 Chr.2: 136116256-136116256 on Build GRCh38
rs964701102 Chr.2: 136116225-136116225 on Build GRCh38
rs958414960 Chr.2: 136116421-136116421 on Build GRCh38
rs993861618 Chr.2: 136116368-136116369 on Build GRCh38
rs569525530 Chr.2: 136116458-136116458 on Build GRCh38
rs575495658 Chr.2: 136116297-136116297 on Build GRCh38
rs560223240 Chr.2: 136116406-136116406 on Build GRCh38
rs2471859 Chr.2: 136116434-136116434 on Build GRCh38
rs897103919 Chr.2: 136116085-136116085 on Build GRCh38
rs923351074 Chr.2: 136116254-136116254 on Build GRCh38
rs949028383 Chr.2: 136116047-136116047 on Build GRCh38
rs753874641 Chr.2: 136116176-136116176 on Build GRCh38
rs990598086 Chr.2: 136116429-136116429 on Build GRCh38

View Full Product Details