Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2133564-2133703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_000612.5 NM_001007139.5 NM_001127598.2 NM_001291861.2 NM_001291862.2 NR_003512.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6978242 c.185G>A p.R62H Chr.11: 2133638-2133638 on Build GRCh38
5495418 c.377G>A p.C126Y Chr.11: 2133614-2133614 on Build GRCh38
6977482 c.182G>A p.R61H Chr.11: 2133641-2133641 on Build GRCh38
6924238 c.166G>A p.A56T Chr.11: 2133657-2133657 on Build GRCh38
6934574 c.396C>A p.D132E Chr.11: 2133595-2133595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762200142 Chr.11: 2133633-2133633 on Build GRCh38
rs773449505 Chr.11: 2133641-2133641 on Build GRCh38
rs369844466 Chr.11: 2133632-2133632 on Build GRCh38
rs150610908 Chr.11: 2133629-2133629 on Build GRCh38
rs774947322 Chr.11: 2133628-2133628 on Build GRCh38
rs754067209 Chr.11: 2133678-2133678 on Build GRCh38
rs745308728 Chr.11: 2133605-2133605 on Build GRCh38
rs1803647 Chr.11: 2133589-2133589 on Build GRCh38
rs768105151 Chr.11: 2133638-2133638 on Build GRCh38
rs377034431 Chr.11: 2133625-2133625 on Build GRCh38
rs761163170 Chr.11: 2133650-2133650 on Build GRCh38
rs113560291 Chr.11: 2133667-2133667 on Build GRCh38
rs199968864 Chr.11: 2133658-2133658 on Build GRCh38
rs112634761 Chr.11: 2133666-2133666 on Build GRCh38

View Full Product Details