Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 136117933-136118194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR4
Entrez Gene ID
7852
Gene Name
C-X-C motif chemokine receptor 4
Gene Aliases
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS
UniGene
Hs.593413
Gene Chromosome Location
Chr.2: 136114349-136118155 on Build GRCh38
Transcripts
NM_003467.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6931629 c.7G>A p.G3R Chr.2: 136118054-136118054 on Build GRCh38
5590437 c.4G>A p.E2K Chr.2: 136118057-136118057 on Build GRCh38
6210518 c.15+1G>A p.? Chr.2: 136118045-136118045 on Build GRCh38
6945449 c.4G>T p.E2* Chr.2: 136118057-136118057 on Build GRCh38
3566845 c.9G>A p.G3G Chr.2: 136118052-136118052 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772461295 Chr.2: 136118065-136118065 on Build GRCh38
rs981436139 Chr.2: 136117985-136117985 on Build GRCh38
rs761720005 Chr.2: 136118071-136118071 on Build GRCh38
rs17848053 Chr.2: 136118162-136118162 on Build GRCh38
rs908818052 Chr.2: 136117984-136117984 on Build GRCh38
rs766208541 Chr.2: 136118084-136118084 on Build GRCh38
rs998506709 Chr.2: 136118165-136118165 on Build GRCh38
rs754447450 Chr.2: 136118089-136118089 on Build GRCh38
rs535221466 Chr.2: 136118122-136118122 on Build GRCh38
rs374847203 Chr.2: 136118072-136118072 on Build GRCh38
rs748544535 Chr.2: 136117996-136117996 on Build GRCh38
rs763008917 Chr.2: 136118076-136118076 on Build GRCh38
rs900000293 Chr.2: 136118070-136118070 on Build GRCh38
rs950047108 Chr.2: 136117970-136117970 on Build GRCh38
rs771337111 Chr.2: 136118026-136118026 on Build GRCh38
rs1050005681 Chr.2: 136118149-136118149 on Build GRCh38
rs372805951 Chr.2: 136118035-136118035 on Build GRCh38
rs774545896 Chr.2: 136118028-136118028 on Build GRCh38
rs890801364 Chr.2: 136118130-136118130 on Build GRCh38
rs932813385 Chr.2: 136118114-136118114 on Build GRCh38
rs898627847 Chr.2: 136118113-136118113 on Build GRCh38
rs941594918 Chr.2: 136117988-136117988 on Build GRCh38
rs200912394 Chr.2: 136118010-136118010 on Build GRCh38
rs767181520 Chr.2: 136118109-136118109 on Build GRCh38
rs555180869 Chr.2: 136118124-136118124 on Build GRCh38
rs751111963 Chr.2: 136118088-136118088 on Build GRCh38
rs113497859 Chr.2: 136118045-136118045 on Build GRCh38
rs778156449 Chr.2: 136118007-136118007 on Build GRCh38
rs917872187 Chr.2: 136117963-136117963 on Build GRCh38
rs983451824 Chr.2: 136117969-136117969 on Build GRCh38

View Full Product Details